Produktname
Sucralose ≥98.0% (HPLC)
Produktnummer
69293
Produktmarke
sigma
CAS-Nummer
56038-13-2
Molekulargewicht
397.63
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| SPECIFIC ROTATION (20/D) | 86.0 ± 2.0 Degrees |
| CONCENTRATION | C=1 IN WATER |
| CARBON CONTENT | 36.25 % (THEORY) |
| HYDROGEN CONTENT | 4.82 % (THEORY) |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |