Produktname
Cys-Gly ≥85% (TLC)
Produktnummer
c0166
Produktmarke
sigma
CAS-Nummer
19246-18-5
Molekulargewicht
178.21
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO OFF- WHITE POWDER |
| SOLUBILITY | CLEAR TO SLIGHTLY HAZY COLORLESS TO YELLOW SOLUTION AT 50 MG/ML IN WATER |
| IR SPECTRUM | CONFORMS TO STRUCTURE |
| CARBON | 28.5 TO 38.9% |
| NITROGEN | 13.3 TO 18.2% |
| PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 85% |
| DOCUMENT # | C0166/09/03/09/4 |