Produktname
Demecolcin -Lösung 10 μg/mL in HBSS, ACF Qualified, BioXtra
Produktnummer
d1925
Produktmarke
sigma
CAS-Nummer
477-30-5
Molekulargewicht
Test | Spezifikation |
|---|---|
| Appearance (Turbidity) | Clear |
| Appearance (Form) | Solution |
| pH | 7.1 - 7.7 |
| Osmolality | 273 - 303 mOs/kg |
| Sterility | Pass |
| Demecolcine Concentration | 8 - 12 µg/ml |