Produktname
Dulbeccos Phosphatgepufferte Kochsalzlösung With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture
Produktnummer
d8662
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Appearance (Turbidity) | Clear |
| Appearance (Form) | Solution |
| pH | 6.9 - 7.1 |
| Osmolality | 275 - 304 mOs/kg |
| Salt Toxicity Test | Pass |
| Cell Line | Cell Line - Cell Types |
| Sterility | Pass |
| Endotoxin Level | <= 1.0 EU/ml |