Produktname
GenElute™-E Single Spin RNA Cleanup Kit Reagents and consumables for organic solvent clean-up from RNA. 10, 50 or 250 purifications.
Produktnummer
ec800
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| Expiry | Report result |
| Void OD A230 | max 0.1 A |
| Eluate OD A230 | max 0.2 A |
| Resin concentration (mL) | < 6.75 mL-8.2 mL |
| Void and Eluate spectrum | No peaks at 210 nm visible |
| pH | 8.5 |