Produktname
Fluoreszeinisothiocyanat–Dextran average Mw 59,000-77,000
Produktnummer
fd70
Produktmarke
sigma
CAS-Nummer
60842-46-8
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Light Yellow to Dark Yellow to Dark Orange |
| APPEARANCE (FORM) | Powder |
| Molecular Weight Mw (GPC-Analysis) | 59000 - 77000 |
| Degree of Substitution | mol FITC/mol of Glucose |