Produktname
Proteinase-K–Agarose aus Tritirachium album lyophilized powder
Produktnummer
p9290
Produktmarke
sigma
CAS-Nummer
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE POWDER |
| ENZYMATIC ACTIVITY | 30 TO 140 UNITS/G SOLID |
| UNIT DEFINITION | ONE UNIT WILL HYDROLYZE UREA DENATURED HEMOGLOBIN TO PRODUCE COLOR EQUIVALENT TO 1.0MICROMOLE OF TYROSINE PER MINUTE AT PH7.5 AT 37DEGC. |
| DOCUMENT # P9290/05/23/07/1 |