Direkt zum Inhalt
Merck
EGF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

EGF human

EGF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(e):
Epidermal Growth Factor, URG, Urogastrone

Products

Produktkategorie
Marke
Biologische Quelle
Farbe
Physikalische Form
Reinheit
Qualitätsstufe
Verfahren