Direkt zum Inhalt
Merck
Malzextrakt-Bouillon NutriSelect® Plus, powder, for yeasts, for molds, pack of 500 g, suitable for microbiology

Malzextrakt-Bouillon

Malzextrakt-Bouillon NutriSelect® Plus, powder, for yeasts, for molds, pack of 500 g, suitable for microbiology
Synonym(e):
Liquid Malt Extract (LME), MEB, Yeast-Malt Extract Broth (YMB)

Products

Produktkategorie
Marke
Biologische Quelle
Farbe
Physikalische Form
Qualitätsstufe
Anwendung
Sterilität
Verfahren