Merck
Milli-Q® IX Reinwassersystem mit E-POD® Abgabevorrichtung input: potable tap water, output: type 2 water (> 5 MΩ·cm), The most advanced pure water system for the production of Elix® quality water at a flow rate of 5 L/h, with E-POD® pure water dispenser.
Milli-Q® IX Reinwassersystem mit E-POD® Abgabevorrichtung
Milli-Q® IX Reinwassersystem mit E-POD® Abgabevorrichtung input: potable tap water, output: type 2 water (> 5 MΩ·cm), The most advanced pure water system for the production of Elix® quality water at a flow rate of 5 L/h, with E-POD® pure water dispenser.
Synonym(e):
Milli-Q® IX 7005 Reinwassersystem mit E-POD® Abgabevorrichtung