Merck
  • Influence of Vitamin E on Electron Microscopic Findings Caused by Pharmacologically Induced Epilepsy.
Influence of Vitamin E on Electron Microscopic Findings Caused by Pharmacologically Induced Epilepsy.
Turkish Neurosurgery (1998)
Farmakolojik, D., and Vitamininin, E. E.