Thin Layer Chromatography Identification Test for Dextromethorphan following European Pharmacopeia Monograph on Silica Gel G 254 Plates
Introduction
Dextromethorphan is a drug of the morphinan class. It is a cough suppressant with sedative, dissociative, and stimulant properties (at higher doses) and it is the active ingredient in many over-the-counter cold and cough medicines.
This application note illustrate how Silica Gel G plates are suitable for the analysis of Dextromethorphan following the European pharmacopeia identification TLC test experimental conditions (version 9.0).
Figure 1.Dextromethorphan Hydrobromide
| TLC Plate | Silica Gel 60G F254 20 x 20 cm | Injection volume | 5 µL |
| Detection | Spray with potassium iodobismuthate solution R2 (Potassium iodobismuthate solution: Suspend 1.7 g of bismuth subnitrate R and 20 g of tartaric acid R in 40 mL of water R. To the suspension add 40 mL of a 400 g/L solution of potassium iodide R and stir for 1 h. Filter. The solution may be kept for several days in brown bottles. Mix immediately before use 5 mL of the stock solution with 15 mL of water. | ||
| Mobile Phase | Concentrated ammonia: methanol : methylene chloride : ethyl acetate : toluene 2:13:10:20:55 (v/v/v/v/v ) | ||
| Development | Over 2/3rd of the plate | ||
| Diluent | Methanol | ||
| Drying | In air | ||
| Standard | Dissolve 25 mg of standard timolol maleate in methanol and dilute to 10 ml with the same solvent | ||
| Sample | Dissolve 25 mg substance to be examined in methanol and dilute to 10 ml with the same solvent. |
Figure 2.TLC plate development
Figure 3.Chromatograms
Figure 4.Spectrum
Figure 5.Chromatographic Data (Standard Solution)
Figure 6.Chromatographic Data (Sample Solution)
| Compound | Rf | |
|---|---|---|
| 1 | Dextromethorphan | 0.29 |
| Compound | Rf | |
|---|---|---|
| 1 | Dextromethorphan | 0.29 |
| STD 1 | 2363 |
| STD 2 | 2347 |
| STD 3 | 2494 |
| STD 4 | 2453 |
| STD 5 | 2461 |
| Mean | 2423.6 |
| Standard Deviation | 64.7 |
| RSD (%) | 2.7 |
- The principal spot in the chromatogram obtained with the test solution is similar in position, colour and size to the principal spot in the chromatogram obtained with reference solution.
- All the spots for the reference solution have a repeatability better than 3% RSD.
- Merck silica gel G plates provides repeatable and reproducible results for Dextromethorphan identification test and comply to all requirements as per European Pharmacopoeia.
Materials
Response not successful: Received status code 500