Merck
Try our products for PFAS Testing
PFAS Pin
Perfluoroalkyl substances (PFAS) are a group of human-made Organofluorine compounds, a class of highly fluorinated substances,  commonly known as “forever chemicals” which means they do not break down in the environment like other chemicals. 
PFAS analysis requires accuracy at every step from sample collection through to analysis!. Our solutions empower researchers trying to better understand the effects of PFAS, as well as regulators and labs focused on providing ongoing exposure testing services.
Complete the form provided below to engage in a discussion about your PFAS requirements with our specialist and be eligible to receive a lab coat pin. Alternatively, seize the opportunity to assess our offerings directly by requesting our PFAS SPE sample pack and PFAS Millex® syringe filters to elevate your PFAS analysis capabilities.
SPE Sample Pack
Supelclean™ ENVI-WAX SPE cartridges for PFAS analysis according to  methods like EPA 533, EPA 1633, ISO 25101 and ISO 21675.
Download PFAS Brochure (PDF)
membrane filter
Try the Millex® syringe filters for your testing application. No detectable PFAS in our PES, Nylon and glass fiber Millex®.
Learn more about Millex®
PFAS Pin - Official Terms & Conditions
*Offer valid in Western Europe only while stocks last. Certain offers may not be valid in all geographic regions; sample quantities may be limited, and additional qualifying restrictions may apply - please contact your account manager for more information. Sigma-Aldrich reserves the right to modify or terminate these or any other promotions at any time in its sole discretion.