Skip to Content
Merck

Skip To

BL3H1F1CC8CD

BLD Pharmatech

4-Aminobenzoic acid

Synonym(s):

4-aminobenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Ships in 14 days - from Aldrich Partner.
€ 48,00
100 g
Ships in 14 days - from Aldrich Partner.
€ 51,00
500 g
Ships in 14 days - from Aldrich Partner.
€ 54,00
1 kg
Ships in 14 days - from Aldrich Partner.
€ 93,00

About This Item

Linear Formula:
C7H7O2N1
CAS Number:
Molecular Weight:
137.14
MDL number:

Vendor SKU: BD17215-25G

€ 48,00


Ships in 14 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

Inchi Key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder

Melting Point

186-189C



Still not finding the right product?

Explore all of our products under 4-Aminobenzoic acid


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library