Skip to Content
Merck

Skip To

BL3H1F1C9471

BLD Pharmatech

4-Amino-1-methylpiperidine

Synonym(s):

1-methyl-4-piperidinylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Ships in 14 days - from Aldrich Partner.
€ 20,00
5 g
Ships in 14 days - from Aldrich Partner.
€ 23,00
10 g
Ships in 14 days - from Aldrich Partner.
€ 26,00
25 g
Ships in 14 days - from Aldrich Partner.
€ 30,00

About This Item

Linear Formula:
C6H14N2
CAS Number:
Molecular Weight:
114.19
MDL number:

Vendor SKU: BD3037-1G

€ 20,00


Ships in 14 days - from Aldrich Partner.

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H14N2/c1-8-4-2-6(7)3-5-8/h6H,2-5,7H2,1H3

Inchi Key

ALOCUZOKRULSAA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?

Explore all of our products under 4-Amino-1-methylpiperidine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library