Skip to Content
Merck

SY3H9A7BD53D

Synthonix Corporation

1-(2-Chloroethyl)piperidine-4-carbonitrile hydrochloride

Sign In to View Organizational & Contract Pricing

Select a Size

Product SY3H9A7BD53D is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
209.12
MDL number:
Product SY3H9A7BD53D is not currently sold in your country.

Iupac Name

1-(2-chloroethyl)-4-piperidinecarbonitrile hydrochloride

Inchi Code

1S/C8H13ClN2.ClH/c9-3-6-11-4-1-8(7-10)2-5-11;/h8H,1-6H2;1H

InChI key

DDMSKIKYADWREH-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/92709.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://synthonix.com/92709.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library