Product Name
Sodium cacodylate Buffer pH 6.5; 50 mM – Isopropanol 15% solution
Product Number
97068
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
DENSITY D20/40.983 - 0.987
REFRACTIVE INDEX N20/D1.344 - 1.346
PH6.3 - 6.7