Skip to Content
Merck
Copper foil, ≥99.8% (complexometric)

Copper

Copper foil, ≥99.8% (complexometric)
Linear Formula:
Cu
CAS Number:
Molecular Weight:
63.55
EC Number:

Products

Brand
Sustainability
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing