Skip to Content
Products
Cart
0
AT
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Phenolphthalein solution 0.375% in methanol
Products
Product Category
indicators
Brand
Supelco
Physical Form
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.07238
Phenolphthalein solution 0.375% in methanol,
Expand
View Pricing
An unknown error has occured.