Merck
  • ARC15105 is a potent antagonist of von Willebrand factor mediated platelet activation and adhesion.
ARC15105 is a potent antagonist of von Willebrand factor mediated platelet activation and adhesion.
Arteriosclerosis, thrombosis, and vascular biology (2012-01-28)
Jolanta M Siller-Matula, Yahye Merhi, Jean-François Tanguay, Daniel Duerschmied, Denisa D Wagner, Kathleen E McGinness, P Shannon Pendergrast, Jou-Ku Chung, Xianbin Tian, Robert G Schaub, Bernd Jilma
ABSTRACT
We investigated the stability, pharmacokinetic, and pharmacodynamic profile of the 2(nd) generation anti-von Willeband factor aptamer ARC15105. Platelet plug formation was measured by collagen/adenosine diphosphate-induced closure time with the platelet function analyzer-100 and platelet aggregation by multiple electrode aggregometry. Platelet adhesion was measured on denuded porcine aortas and in a flow chamber. Aptamer stability was assessed by incubation in nuclease rich human, monkey, and rat serum for up to 72 hours. Pharmacokinetic and pharmacodynamic profiles were tested in cynomolgus monkeys after IV and SC administration. The median IC(100) and IC(50) to prolong collagen/adenosine diphosphate-induced closure timewere 27 nmol/L and 12 nmol/L, respectively. ARC15105 (1.3 μmol/L) completely inhibited ristocetin-induced platelet aggregation in whole blood (P<0.001), but also diminished collagen, ADP, arachidonic acid, and thrombin receptor activating peptide-induced platelet aggregation to some extent (P<0.05). ARC15105 (40 nmol/L) decreased platelet adhesion by >90% on denuded porcine aortas (P<0.001), which was comparable to the degree of inhibition obtained with abciximab. ARC15105 (100 nmol/L) also inhibited platelet adhesion to collagen under arterial shear in a flow chamber by >90% (P<0.001). The IV and SC terminal half-lives in cynomolgus monkeys were 67 and 65 hours, respectively, and the SC bioavailability was ≈98%. Allometric scaling estimates the human T(1/2) would be ≈217 hours. Pharmacodynamic analysis confirms that ARC15105 inhibits von Willeband factor activity >90% in blood samples taken 300 hours after a 20 mg/kg IV or SC dose in monkeys. The potency, pharmacokinetic profile, and SC bioavailability of ARC15105 support its clinical investigation for chronic inhibition of von Willeband factor -mediated platelet activation.