Merck
  • Computerised morphometrical analysis in endometrial hyperplasia for the prediction of cancer development. A long-term retrospective study from northern Norway.
Computerised morphometrical analysis in endometrial hyperplasia for the prediction of cancer development. A long-term retrospective study from northern Norway.
Journal of clinical pathology (2000-10-21)
A Orbo, J P Baak, I Kleivan, S Lysne, P S Prytz, M A Broeckaert, A Slappendel, H J Tichelaar
ABSTRACT
To evaluate and compare the long term prognostic value of the WHO classification and the computerised multivariate morphometrical D score in endometrial hyperplasia. To test the reproducibility of the D score in two different centres. Histopathological WHO classification and computerised morphometrical analysis using the D score (< 0, high risk; > 1, low risk; 0-1, uncertain) in a population based study from northern Norway of archival dilatation and curettage material from 68 women with 10-20 years of follow up. Of the 68 patients included in the study, 18 developed cancer. The sensitivity and specificity of the D score (< 0 v > 1) were 100% and 78%, respectively, which was better than the WHO classification (89% and 60%, respectively). The negative and positive predictive values for the D score were 100% and 58% and of the WHO classification 94% and 44%, respectively. This study found a slightly higher specificity for the D score than former retrospective studies, but otherwise the results were comparable. The D score results were reproducible between the two centres (R = 0.91; slope = 0.98; intercept = 0.3). D score assessment is a reproducible and more accurate predictor of outcome of endometrial hyperplasia than the WHO classification assessed by an experienced gynaecological pathologist. Routine application of the D score might reduce over and undertreatment of endometrial hyperplasia.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea