Merck
  • Dietary tetrahydrocurcumin reduces renal fibrosis and cardiac hypertrophy in 5/6 nephrectomized rats.
Dietary tetrahydrocurcumin reduces renal fibrosis and cardiac hypertrophy in 5/6 nephrectomized rats.
Pharmacology research & perspectives (2018-02-23)
Wei Ling Lau, Mahyar Khazaeli, Javad Savoj, Kasim Manekia, Maria Bangash, Roshni G Thakurta, Anhthu Dang, Nosratola D Vaziri, Bhupinder Singh
ABSTRACT
Tetrahydrocurcumin (THC) is the principal metabolite of curcumin and has antioxidant properties. In the present investigation, the effect of THC on renal and cardiovascular outcomes was studied in rats with chronic kidney disease (CKD). CKD rats were randomized following 5/6 nephrectomy to a special diet for 9 weeks which contained 1% THC (CKD+THC group). Low-dose polyenylphosphatidylcholine was used as a lipid carrier to increase bioavailability. Endpoints included tail blood pressure, normalized heart weight, plasma and urine biochemical data, and kidney tissue analyses. CKD animals demonstrated increased proteinuria, decreased creatinine clearance, hypertension, and cardiac hypertrophy. The antioxidant proteins CuZn SOD and glutathione peroxidase were decreased in the remnant kidney, while apoptosis (caspase-3) and fibrosis (alpha-SM actin) were increased. Renal fibrosis was confirmed histologically on trichrome staining. These pathologic changes were ameliorated in the CKD+THC group with significant decrease in proteinuria, hypertension, and kidney fibrosis. THC therapy restored levels of CuZn SOD and glutathione peroxidase. Consistent with prior reports, dietary THC did not improve nuclear Nrf2 levels. In summary, dietary THC therapy improved expression of antioxidant proteins in the remnant kidney, decreased renal fibrosis and proteinuria, and ameliorated hypertension in 5/6 nephrectomized rats.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NRF2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Catalase antibody ,Mouse monoclonal, clone CAT-505, purified from hybridoma cell culture