Merck
  • Amoeboid microglia in the periventricular white matter induce oligodendrocyte damage through expression of proinflammatory cytokines via MAP kinase signaling pathway in hypoxic neonatal rats.
Amoeboid microglia in the periventricular white matter induce oligodendrocyte damage through expression of proinflammatory cytokines via MAP kinase signaling pathway in hypoxic neonatal rats.
Brain pathology (Zurich, Switzerland) (2008-03-29)
YiYu Deng, Jia Lu, Viswanathan Sivakumar, Eng Ang Ling, Charanjit Kaur
ABSTRACT
Hypoxic injury in the perinatal period results in periventricular white matter (PWM) lesions with axonal damage and oligodendroglial loss. It also alters macrophage function by perpetuating expression of inflammatory mediators. Relevant to this is the preponderance of amoeboid microglial cells (AMC) characterized as active macrophages in the developing PWM. This study aimed to determine if AMC produce proinflammatory cytokines that may be linked to the oligodendroglial loss observed in hypoxic PWM damage (PWMD). Wistar rats (1 day old) were subjected to hypoxia, following which upregulated expression of tumor necrosis factor-alpha (TNF-alpha), interleukin-1beta (IL-1beta), TNF receptor 1 (TNF-R(1)) and IL-1 receptor 1 (IL-1R(1)) was observed. This was coupled with apoptosis and expression of TNF-R(1) and IL-1R(1) in oligodendrocytes. Primary cultured microglial cells subjected to hypoxia (3% oxygen, 5% CO(2) and 92% nitrogen) showed enhanced expression of TNF-alpha and IL-1beta. Furthermore, mitogen-activated protein (MAP) kinase signaling pathway was involved in the expression of TNF-alpha and IL-1beta in microglia subjected to hypoxia. Our results suggest that following a hypoxic insult, microglial cells in the neonatal rats produce inflammatory cytokines such as TNF-alpha and IL-1beta via MAP kinase signaling pathway. These cytokines are detrimental to oligodendrocytes resulting in PWM lesion.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Mouse IgG & IgM Antibody, FITC conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Actin (α-Sarcomeric) antibody produced in mouse, clone 5C5, ascites fluid
Sigma-Aldrich
Anti-Interleukin-1β Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Tumor Necrosis Factor-α Antibody, Chemicon®, from rabbit