- Stats: transcriptional control and biological impact.
Stats: transcriptional control and biological impact.
Nature reviews. Molecular cell biology (2002-09-05)
David E Levy, J E Darnell
PMID12209125
ABSTRACT
Extracellular proteins bound to cell-surface receptors can change nuclear gene expression patterns in minutes, with far-reaching consequences for development, cell growth and homeostasis. The signal transducer and activator of transcription (STAT) proteins are among the most well studied of the latent cytoplasmic signal-dependent transcription-factor pathways. In addition to several roles in normal cell decisions, dysregulation of STAT function contributes to human disease, making the study of these proteins an important topic of current research.