Merck
  • MCP-1 is overexpressed in triple-negative breast cancers and drives cancer invasiveness and metastasis.
MCP-1 is overexpressed in triple-negative breast cancers and drives cancer invasiveness and metastasis.
Breast cancer research and treatment (2018-03-30)
Pranabananda Dutta, Marianna Sarkissyan, Kimberly Paico, Yanyuan Wu, Jaydutt V Vadgama
ABSTRACT
Triple-negative breast cancer (TNBC) is the most aggressive type of breast cancer that lacks ER/PR and HER2 receptors. Hence, there is urgency in developing new or novel therapeutic strategies for treatment of TNBC. Our study shows that the Monocyte Chemoattractant Protein-1 (MCP-1) is a marker associated with TNBC and may play a key role in TNBC disease progression. ELISA method was used to measure secreted MCP-1, and mRNA levels were determined by Real-time PCR in numerous cancer cell lines, representing various breast cancer subtypes. Cellular invasiveness was determined by Boyden chamber assay. Our data show that MCP-1 is upregulated in TNBC cell lines both transcriptionally as well as in secreted protein levels compared to ER-positive luminal cell line, MCF-7. Breast cancer patients, with Basal or Claudin-low subtypes, also showed high expression of MCP-1. MCP-1 treatment induced cell invasion in various breast cancer cell types, without affecting cell proliferation. Small molecule antagonists against Chemokine Receptor 2 (CCR2), cognate receptor for MCP-1 as well as the MAP kinase pathway inhibitor U0126 negatively affected MCP-1 induced MCF-7 cell invasion. This suggests that MCP-1-CCR2 axis may regulate invasiveness via the MAP Kinase pathway. Knocking down MCP-1 decreased cell invasion in TNBC cell line BT-549, along with downregulation of key epithelial to mesenchymal transition markers, N-cadherin and Vimentin. Our study suggests that MCP-1 mediated pathways could be potential therapeutic targets for the treatment of TNBC, and could reduce cancer health disparities.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCL2