Merck
Rapid Cue-Specific Remodeling of the Nascent Axonal Proteome.
Neuron (2018-07-17)
Roberta Cagnetta, Christian K Frese, Toshiaki Shigeoka, Jeroen Krijgsveld, Christine E Holt
ABSTRACT
Axonal protein synthesis and degradation are rapidly regulated by extrinsic signals during neural wiring, but the full landscape of proteomic changes remains unknown due to limitations in axon sampling and sensitivity. By combining pulsed stable isotope labeling of amino acids in cell culture with single-pot solid-phase-enhanced sample preparation, we characterized the nascent proteome of isolated retinal axons on an unparalleled rapid timescale (5 min). Our analysis detects 350 basally translated axonal proteins on average, including several linked to neurological disease. Axons stimulated by different cues (Netrin-1, BDNF, Sema3A) show distinct signatures with more than 100 different nascent protein species up- or downregulated within the first 5 min followed by further dynamic remodeling. Switching repulsion to attraction triggers opposite regulation of a subset of common nascent proteins. Our findings thus reveal the rapid remodeling of the axonal proteomic landscape by extrinsic cues and uncover a logic underlying attraction versus repulsion.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000 by viscosity
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)
Sigma-Aldrich
Anti-Protein Disulfide Isomerase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
ANTI-HIST1H2BL (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin γ Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse