Merck
  • Potent inhibitory action of chlorethylclonidine on the positive inotropic effect and phosphoinositide hydrolysis mediated via myocardial alpha 1-adrenoceptors in the rabbit ventricular myocardium.
Potent inhibitory action of chlorethylclonidine on the positive inotropic effect and phosphoinositide hydrolysis mediated via myocardial alpha 1-adrenoceptors in the rabbit ventricular myocardium.
Naunyn-Schmiedeberg's archives of pharmacology (1991-06-01)
M Takanashi, I Norota, M Endoh
ABSTRACT
The influence of the alpha 1b-adrenoceptor-selective antagonist chlorethylclonidine on the alpha 1-adrenergic positive inotropic effect and the phosphoinositide hydrolysis induced by phenylephrine was investigated in the rabbit ventricular myocardium. Pretreatment of membrane fractions derived from the rabbit ventricular muscle with 10(-5) mol/l chlorethylclonidine decreased the specific binding of [3H]prazosin (at a saturating concentration of 10(-9) mol/l) from the control value of 11.27 +/- 0.48 to 4.18 +/- 1.87 fmol/mg protein. The inhibition by adrenaline of the binding of [3H]prazosin (slope factor and affinity) was not affected by chlorethylclonidine. The positive inotropic effect of phenylephrine (in the presence of 3 x 10(-7) mol/l bupranolol) was inhibited by chlorethylclonidine in a concentration-dependent manner (10(-7)-10(-5) mol/l) and abolished by 10(-5) mol/l chlorethylclonidine. The concentration of chlorethylclonidine to inhibit the phenylephrine-induced maximum response to 50% was 2.4 x 10(-6) mol/l. The accumulation of [3H]inositol monophosphate and [3H]inositol trisphosphate induced by 10(-5) mol/l phenylephrine was inhibited by chlorethylclonidine in the same concentration range. These findings indicate that the myocardial alpha 1-adrenoceptors mediating a positive inotropic effect in the rabbit ventricular myocardium may belong to the chlorethylclonidine-sensitive alpha 1b-subtype, and that the subcellular mechanism of action involve phosphoinositide hydrolysis.