Merck
  • LIM and cysteine-rich domains 1 is required for thrombin-induced smooth muscle cell proliferation and promotes atherogenesis.
LIM and cysteine-rich domains 1 is required for thrombin-induced smooth muscle cell proliferation and promotes atherogenesis.
The Journal of biological chemistry (2018-01-13)
Jagadeesh Janjanam, Baolin Zhang, Arul M Mani, Nikhlesh K Singh, James G Traylor, A Wayne Orr, Gadiparthi N Rao
ABSTRACT
Restenosis arises after vascular injury and is characterized by arterial wall thickening and decreased arterial lumen space. Vascular injury induces the production of thrombin, which in addition to its role in blood clotting acts as a mitogenic and chemotactic factor. In exploring the molecular mechanisms underlying restenosis, here we identified LMCD1 (LIM and cysteine-rich domains 1) as a gene highly responsive to thrombin in human aortic smooth muscle cells (HASMCs). Of note, LMCD1 depletion inhibited proliferation of human but not murine vascular smooth muscle cells. We also found that by physically interacting with E2F transcription factor 1, LMCD1 mediates thrombin-induced expression of the CDC6 (cell division cycle 6) gene in the stimulation of HASMC proliferation. Thrombin-induced LMCD1 and CDC6 expression exhibited a requirement for protease-activated receptor 1-mediated Gαq/11-dependent activation of phospholipase C β3. Moreover, the expression of LMCD1 was highly induced in smooth muscle cells located at human atherosclerotic lesions and correlated with CDC6 expression and that of the proliferation marker Ki67. Furthermore, the LMCD1- and SMCαactin-positive cells had higher cholesterol levels in the atherosclerotic lesions. In conclusion, these findings indicate that by acting as a co-activator with E2F transcription factor 1 in CDC6 expression, LMCD1 stimulates HASMC proliferation and thereby promotes human atherogenesis, suggesting an involvement of LMCD1 in restenosis.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit