Merck
  • Assessing the corrosion of biodegradable magnesium implants: a critical review of current methodologies and their limitations.
Assessing the corrosion of biodegradable magnesium implants: a critical review of current methodologies and their limitations.
Acta Biomaterialia (2012)
Kirkland Nt