- Increasing the Receptor Tyrosine Kinase EphB2 Prevents Amyloid-?-induced Depletion of Cell Surface Glutamate Receptors by a Mechanism That Requires the PDZ-binding Motif of EphB2 and Neuronal Activity
Increasing the Receptor Tyrosine Kinase EphB2 Prevents Amyloid-?-induced Depletion of Cell Surface Glutamate Receptors by a Mechanism That Requires the PDZ-binding Motif of EphB2 and Neuronal Activity
The Journal of Biological Chemistry (2015)
Miyamoto T, et al.