Merck
  • PD-1 Blockade for Improving the Antitumor Efficiency of Polymer-Doxorubicin Nanoprodrug.
PD-1 Blockade for Improving the Antitumor Efficiency of Polymer-Doxorubicin Nanoprodrug.
Small (Weinheim an der Bergstrasse, Germany) (2018-08-22)
Fan Gao, Chi Zhang, Wen-Xiu Qiu, Xue Dong, Di-Wei Zheng, Wei Wu, Xian-Zheng Zhang
ABSTRACT
Chemotherapy is well recognized to induce immune responses during some chemotherapeutic drugs-mediated tumor eradication. Here, a strategy involving blocking programmed cell death protein 1 (PD-1) to enhance the chemotherapeutic effect of a doxorubicin nanoprodrug HA-Psi-DOX is proposed and the synergetic mechanism between them is further studied. The nanoprodrugs are fabricated by conjugating doxorubicin (DOX) to an anionic polymer hyaluronic acid (HA) via a tumor overexpressed matrix metalloproteinase sensitive peptide (CPLGLAGG) for tumor targeting and enzyme-activated drug release. Once accumulated at the tumor site, the nanoprodrug can be activated to release antitumor drug by tumor overexpressed MMP-2. It is found that HA-Psi-DOX nanoparticles can kill tumor cells effectively and initiate an antitumor immune response, leading to the upregulation of interferon-γ. This cytokine promotes the expression of programmed cell death protein-ligand 1 (PD-L1) on tumor cells, which will cause immunosuppression after interacting with PD-1 on the surface of lymphocytes. The results suggest that the therapeutic efficiency of HA-Psi-DOX nanoparticles is significantly improved when combined with checkpoint inhibitors anti-PD-1 antibody (α-PD1) due to the neutralization of immunosuppression by blocking the interaction between PD-L1 and PD-1. This therapeutic system by combining chemotherapy and immunotherapy further increases the link between conventional tumor therapies and immunotherapy.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Fmoc-Ala-OH, 95%
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Fmoc-Leu-OH, ≥97.0%
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)