Merck
  • Accumulation of 8,9-unsaturated sterols drives oligodendrocyte formation and remyelination.
Accumulation of 8,9-unsaturated sterols drives oligodendrocyte formation and remyelination.
Nature (2018-07-27)
Zita Hubler, Dharmaraja Allimuthu, Ilya Bederman, Matthew S Elitt, Mayur Madhavan, Kevin C Allan, H Elizabeth Shick, Eric Garrison, Molly T Karl, Daniel C Factor, Zachary S Nevin, Joel L Sax, Matthew A Thompson, Yuriy Fedorov, Jing Jin, William K Wilson, Martin Giera, Franz Bracher, Robert H Miller, Paul J Tesar, Drew J Adams
ABSTRACT
Regeneration of myelin is mediated by oligodendrocyte progenitor cells-an abundant stem cell population in the central nervous system (CNS) and the principal source of new myelinating oligodendrocytes. Loss of myelin-producing oligodendrocytes in the CNS underlies a number of neurological diseases, including multiple sclerosis and diverse genetic diseases1-3. High-throughput chemical screening approaches have been used to identify small molecules that stimulate the formation of oligodendrocytes from oligodendrocyte progenitor cells and functionally enhance remyelination in vivo4-10. Here we show that a wide range of these pro-myelinating small molecules function not through their canonical targets but by directly inhibiting CYP51, TM7SF2, or EBP, a narrow range of enzymes within the cholesterol biosynthesis pathway. Subsequent accumulation of the 8,9-unsaturated sterol substrates of these enzymes is a key mechanistic node that promotes oligodendrocyte formation, as 8,9-unsaturated sterols are effective when supplied to oligodendrocyte progenitor cells in purified form whereas analogous sterols that lack this structural feature have no effect. Collectively, our results define a unifying sterol-based mechanism of action for most known small-molecule enhancers of oligodendrocyte formation and highlight specific targets to propel the development of optimal remyelinating therapeutics.
Sigma-Aldrich
Desmosterol, ≥84% (GC)
Sigma-Aldrich
Lanosterol, ≥93%, powder
Sigma-Aldrich
UNC1999, ≥98% (HPLC)
Sigma-Aldrich
Bexarotene, ≥98% (HPLC)
Sigma-Aldrich
GSK343, ≥98% (HPLC)
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Anti-NG2, Alexa Fluor488 Conjugate Antibody, from rabbit, ALEXA FLUOR 488