Merck
  • OnabotulinumtoxinA for the Treatment of Poststroke Distal Lower Limb Spasticity: A Randomized Trial.
OnabotulinumtoxinA for the Treatment of Poststroke Distal Lower Limb Spasticity: A Randomized Trial.
PM & R : the journal of injury, function, and rehabilitation (2018-01-14)
Theodore Wein, Alberto Esquenazi, Wolfgang H Jost, Anthony B Ward, Grace Pan, Rozalina Dimitrova
ABSTRACT
Poststroke distal lower limb spasticity impairs mobility, limiting activities of daily living and requiring additional caregiver time. To evaluate the efficacy, safety, and sustained benefit of onabotulinumtoxinA in adults with poststroke lower limb spasticity (PSLLS). A multicenter, randomized, double-blind, phase 3, placebo-controlled trial (NCT01575054). Sixty study centers across North America, Europe, Russia, the United Kingdom, and South Korea. Adult patients (18-65 years of age) with PSLLS (Modified Ashworth Scale [MAS] ≥3) of the ankle plantar flexors and the most recent stroke ≥3 months before study enrollment. During the open-label phase, patients received ≤3 onabotulinumtoxinA treatments (≤400 U) or placebo at approximately 12-week intervals. Treatments were into the ankle plantar flexors (onabotulinumtoxinA 300 U into ankle plantar flexors; ≤100 U, optional lower limb muscles). The double-blind primary endpoint was MAS change from baseline (average score at weeks 4 and 6). Secondary measures included physician-assessed Clinical Global Impression of Change (CGI), MAS change from baseline in optional muscles, Goal Attainment Scale (GAS), and pain scale. Of 468 patients enrolled, 450 (96%) completed the double-blind phase and 413 (88%) completed the study. Small improvements in MAS observed with onabotulinumtoxinA during the double-blind phase (onabotulinumtoxinA, -0.8; placebo, -0.6, P = .01) were further enhanced with additional treatments through week 6 of the third open-label treatment cycle (onabotulinumtoxinA/onabotulinumtoxinA, -1.2; placebo/onabotulinumtoxinA, -1.4). Small improvements in CGI observed during the double-blind phase (onabotulinumtoxinA, 0.9; placebo, 0.7, P = .01) were also further enhanced through week 6 of the third open-label treatment cycle (onabotulinumtoxinA/onabotulinumtoxinA, 1.6; placebo/onabotulinumtoxinA, 1.6). Physician- and patient-assessed GAS scores improved with each subsequent treatment. No new safety signals emerged. OnabotulinumtoxinA significantly improved ankle MAS, CGI, and GAS scores compared with placebo; improvements were consistent and increased with repeated treatments of onabotulinumtoxinA over 1 year in patients with PSLLS. I.
Sigma-Aldrich
Allyl methyl sulfone, 96%