Merck
  • Isolation and characterization of a native placental basement-membrane collagen and its component alpha chains.
Isolation and characterization of a native placental basement-membrane collagen and its component alpha chains.
European journal of biochemistry (1979-04-02)
R W Glanville, A Rauter, P P Fietzek
PMID456357
ABSTRACT
Native type IV collagen was isolated from human placenta using pepsin solubilisation followed by fractional salt precipitation and chromatogarphic purification. The native preparation was characterised using amino acid analyses, disc gel electrophoresis, segment-long-spacing crystallites and immunological methods. Two component alpha chains were isolated with molecular weights of approximately 95000 and 70000. Cyanogen bromide digests of these chains indicated that they are not related to any of the known alpha chains of interstitial collagens or to the recently described collagen containing alphaA and alphaB chains. They are also not related to one another and are therefore probably fragments of two genetically distinct type IV collagen alpha chains.
Sigma-Aldrich
Human Collagen Type IV, 1 mg/mL, solution, BioReagent, suitable for cell culture
Sigma-Aldrich
Human Collagen Type I, 1 mg/mL, solution, BioReagent, suitable for cell culture
Sigma-Aldrich
Human Collagen Type V, 1 mg/mL, solution, BioReagent, suitable for cell culture
Sigma-Aldrich
Human Collagen Type III, 1 mg/mL, solution, Bioreagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder