Merck
  • MicroRNA-27a controls the intracellular survival of Mycobacterium tuberculosis by regulating calcium-associated autophagy.
MicroRNA-27a controls the intracellular survival of Mycobacterium tuberculosis by regulating calcium-associated autophagy.
Nature communications (2018-10-18)
Feng Liu, Jianxia Chen, Peng Wang, Haohao Li, Yilong Zhou, Haipeng Liu, Zhonghua Liu, Ruijuan Zheng, Lin Wang, Hua Yang, Zhenling Cui, Fei Wang, Xiaochen Huang, Jie Wang, Wei Sha, Heping Xiao, Baoxue Ge
ABSTRACT
Tuberculosis (TB) caused by Mycobacterium tuberculosis (Mtb) kills millions every year, and there is urgent need to develop novel anti-TB agents due to the fast-growing of drug-resistant TB. Although autophagy regulates the intracellular survival of Mtb, the role of calcium (Ca2+) signaling in modulating autophagy during Mtb infection remains largely unknown. Here, we show that microRNA miR-27a is abundantly expressed in active TB patients, Mtb-infected mice and macrophages. The target of miR-27a is the ER-located Ca2+ transporter CACNA2D3. Targeting of this transporter leads to the downregulation of Ca2+ signaling, thus inhibiting autophagosome formation and promoting the intracellular survival of Mtb. Mice lacking of miR-27a and mice treated with an antagomir to miR-27a are more resistant to Mtb infection. Our findings reveal a strategy for Mtb to increase intracellular survival by manipulating the Ca2+-associated autophagy, and may also support the development of host-directed anti-TB therapeutic approaches.
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Pricing and availability is not currently available.
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Pricing and availability is not currently available.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).
Pricing and availability is not currently available.
Sigma-Aldrich
LookOut® Mycoplasma PCR Detection Kit, Optimized for use with JumpStart Taq DNA Polymerase, D9307.
Pricing and availability is not currently available.
Sigma-Aldrich
1-Pyrrolidinecarbodithioic Acid, Ammonium Salt, Cell permeable. Inhibits the induction of NOS activity in rat alveolar macrophages. Prevents apoptosis in human HL-60 cells and thymocytes, but induces apoptosis in human and rat smooth muscle cells.
Pricing and availability is not currently available.