Merck
  • Binding of [Cr (phen) 3] 3+ to transferrin at extracellular and endosomal pHs: Potential application in photodynamic therapy
Binding of [Cr (phen) 3] 3+ to transferrin at extracellular and endosomal pHs: Potential application in photodynamic therapy
Biochim. Biophys. Acta Gen. Subj. (2014)
Garcia PF, et al.