Merck
  • Fine depth resolution of two-photon absorption-induced photoacoustic microscopy using low-frequency bandpass filtering
Fine depth resolution of two-photon absorption-induced photoacoustic microscopy using low-frequency bandpass filtering
Optics Express (2011)
YamaokaY, et al.