- Curcumin effectively rescued Parkinson?s disease-like phenotypes in a novel drosophila melanogaster model with dUCH Knockdown
Curcumin effectively rescued Parkinson?s disease-like phenotypes in a novel drosophila melanogaster model with dUCH Knockdown
Oxidative Medicine and Cellular Longevity (2018)
Nguyen TT, et al.