Merck
  • Hyperinsulinemia and insulin resistance in Wrn null mice fed a diabetogenic diet.
Hyperinsulinemia and insulin resistance in Wrn null mice fed a diabetogenic diet.
Mechanisms of ageing and development (2008-02-26)
Gina Moore, Susan Knoblaugh, Kathryn Gollahon, Peter Rabinovitch, Warren Ladiges
ABSTRACT
Werner syndrome (WS) is an autosomal recessive progeroid syndrome caused by mutations in the Werner (Wrn) gene. WS patients have increased incidence of a number of chronic conditions including insulin resistance and type 2 diabetes. Since ingestion of foods that are high in fat and sugar is associated with increased incidence of diabetes, we examined if Wrn mutations might affect metabolic response to a diabetogenic diet. Four-month-old mice with a null mutation for the Wrn gene were fed a diet consisting of 36% fat (lard), 33% table sugar, and 20% protein plus balanced vitamins and minerals. Wrn null mice had significantly increased body weights, increased serum insulin levels, impaired glucose tolerance, and insulin resistance during 4 months of eating the diabetogenic diet. Diffuse fatty infiltration of the liver and pancreatic islet hyperplasia was characteristic morphological features. These observations suggest that Wrn null mice have impaired glucose homeostasis and fat metabolism, and may be a useful model to investigate metabolic conditions associated with aging.
Product Description
Millipore
Rat/Mouse Insulin ELISA, This Rat/Mouse Insulin ELISA is used to measure & quantify Insulin levels in Metabolism & Endocrine research.
Millipore
Mouse Leptin ELISA, This Mouse Leptin ELISA is used to measure & quantify Leptin levels in Metabolism & Endocrine & Neuroscience research.
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Serum Triglyceride Determination Kit, 1 kit sufficient for 250 tests