Merck
A revised structure of fluorescein mercuric acetate.
Analytical biochemistry (1986-05-01)
M T Shipchandler, J R Fino
ABSTRACT
With the help of NMR spectroscopy the structure assignment of fluorescein mercuric acetate is corrected to 4',5'-bis(acetoxymercury)fluorescein.