Merck
  • Aberrant hippocampal neurogenesis contributes to epilepsy and associated cognitive decline.
Aberrant hippocampal neurogenesis contributes to epilepsy and associated cognitive decline.
Nature communications (2015-03-27)
Kyung-Ok Cho, Zane R Lybrand, Naoki Ito, Rebecca Brulet, Farrah Tafacory, Ling Zhang, Levi Good, Kerstin Ure, Steven G Kernie, Shari G Birnbaum, Helen E Scharfman, Amelia J Eisch, Jenny Hsieh
ABSTRACT
Acute seizures after a severe brain insult can often lead to epilepsy and cognitive impairment. Aberrant hippocampal neurogenesis follows the insult but the role of adult-generated neurons in the development of chronic seizures or associated cognitive deficits remains to be determined. Here we show that the ablation of adult neurogenesis before pilocarpine-induced acute seizures in mice leads to a reduction in chronic seizure frequency. We also show that ablation of neurogenesis normalizes epilepsy-associated cognitive deficits. Remarkably, the effect of ablating adult neurogenesis before acute seizures is long lasting as it suppresses chronic seizure frequency for nearly 1 year. These findings establish a key role of neurogenesis in chronic seizure development and associated memory impairment and suggest that targeting aberrant hippocampal neurogenesis may reduce recurrent seizures and restore cognitive function following a pro-epileptic brain insult.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
3-Methylpyridine, ≥99.5%
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC
Sigma-Aldrich
Anti-Prox 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Doublecortin Antibody, serum, from guinea pig