Merck
  • A continuous fluorometric assay for trypsin based on melittin and the noncovalent-binding-induced pyrene excimer
A continuous fluorometric assay for trypsin based on melittin and the noncovalent-binding-induced pyrene excimer
Chemistry Letters (Jpn) (2013)
Xu N, et al.