Merck
  • Fragmentation patterns of methyloxime-trimethylsilyl derivatives of constitutive mono-and disaccharide isomers analyzed by gas chromatography/field ionization mass spectrometry
Fragmentation patterns of methyloxime-trimethylsilyl derivatives of constitutive mono-and disaccharide isomers analyzed by gas chromatography/field ionization mass spectrometry
Rapid Communications in Mass Spectrometry (2015)
Furuhashi T, et al.