- Nanoparticles containing allotropes of carbon have genotoxic effects on glioblastoma multiforme cells
Nanoparticles containing allotropes of carbon have genotoxic effects on glioblastoma multiforme cells
International journal of nanomedicine (2014)
Hinzmann M, et al.