Merck
  • L-ornithine supplementation attenuates physical fatigue in healthy volunteers by modulating lipid and amino acid metabolism
L-ornithine supplementation attenuates physical fatigue in healthy volunteers by modulating lipid and amino acid metabolism
Nutrition Research (New York, N.Y.) (2008)
Sugino T, et al.