Merck
  • Effect of cholesteryl hemisuccinate on the interfacial properties of phosphatidylcholine bilayers
Effect of cholesteryl hemisuccinate on the interfacial properties of phosphatidylcholine bilayers
Biochimica et Biophysica Acta - Biomembranes (1998)
Massey JB