Merck
  • IL-6/JAK1 pathway drives PD-L1 Y112 phosphorylation to promote cancer immune evasion.
IL-6/JAK1 pathway drives PD-L1 Y112 phosphorylation to promote cancer immune evasion.
The Journal of clinical investigation (2019-07-16)
Li-Chuan Chan, Chia-Wei Li, Weiya Xia, Jung-Mao Hsu, Heng-Huan Lee, Jong-Ho Cha, Hung-Ling Wang, Wen-Hao Yang, Er-Yen Yen, Wei-Chao Chang, Zhengyu Zha, Seung-Oe Lim, Yun-Ju Lai, Chunxiao Liu, Jielin Liu, Qiongzhu Dong, Yi Yang, Linlin Sun, Yongkun Wei, Lei Nie, Jennifer L Hsu, Hui Li, Qinghai Ye, Manal M Hassan, Hesham M Amin, Ahmed O Kaseb, Xin Lin, Shao-Chun Wang, Mien-Chie Hung
ABSTRACT
Glycosylation of immune receptors and ligands, such as T cell receptor and coinhibitory molecules, regulates immune signaling activation and immune surveillance. However, how oncogenic signaling initiates glycosylation of coinhibitory molecules to induce immunosuppression remains unclear. Here we show that IL-6-activated JAK1 phosphorylates programmed death-ligand 1 (PD-L1) Tyr112, which recruits the endoplasmic reticulum-associated N-glycosyltransferase STT3A to catalyze PD-L1 glycosylation and maintain PD-L1 stability. Targeting of IL-6 by IL-6 antibody induced synergistic T cell killing effects when combined with anti-T cell immunoglobulin mucin-3 (anti-Tim-3) therapy in animal models. A positive correlation between IL-6 and PD-L1 expression was also observed in hepatocellular carcinoma patient tumor tissues. These results identify a mechanism regulating PD-L1 glycosylation initiation and suggest the combination of anti-IL-6 and anti-Tim-3 as an effective marker-guided therapeutic strategy.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution