Merck
  • Phosphorylcholine-based hydrogel for immobilization of biomolecules. Application to fluorometric microarrays for use in hybridization assays and immunoassays, and nanophotonic biosensing.
Phosphorylcholine-based hydrogel for immobilization of biomolecules. Application to fluorometric microarrays for use in hybridization assays and immunoassays, and nanophotonic biosensing.
Mikrochimica acta (2019-07-25)
Zeneida Díaz-Betancor, María-José Bañuls, Francisco Javier Sanza, Rafael Casquel, Maria Fe Laguna, Miguel Holgado, Rosa Puchades, Ángel Maquieira
ABSTRACT
An approach is presented for covalent immobilization of biomolecules on an acrylate phosphorylcholine hydrogel. The immobilization and the hydrogel formation take place simultaneously by a thiol-acrylate coupling reaction, induced by UV-light (254 nm). The hydrogel is prepared on two polymeric surfaces (the HardCoat protective layer of Blu-Ray discs, and SU-8) and applied to fluorescence microarray and label-free interferometric detection. For the first, Cy5 labeled analytes are used (λem 635 nm) and, for the second, a periodic array of high-aspect ratio nanopillars detects unlabeled analytes by interferometry. Bioavailability of the immobilized probes is demonstrated in labeled assays; for the case of oligonucleotides by discriminating single nucleotide polymorphisms, and, for the case of antibodies, by BSA immunorecognition. The raw hydrogel is employed to detect human C-reactive protein, in both labeled and non-labeled assay formats, with sensitivities of 30 ng·mL-1 and 2 pg·mL-1, respectively. Graphical abstract Schematic presentation of the phosphorylcholine (MPC) hydrogel preparation onto BluRay disc and SU-8 nanopillars to perform fluorescence and label-free interferometric detection, respectively. It selectively detects C-reactive protein (CRP), but it can covalently immobilize antibodies or nucleid acid probes to detect other analytes.