Merck
  • Validation of simultaneous volumetric and spectrophotometric methods for the determination of captopril in pharmaceutical formulations
Validation of simultaneous volumetric and spectrophotometric methods for the determination of captopril in pharmaceutical formulations
Il Farmaco (Societa Chimica Italiana : 1989) (2005)
Rahman N, et al.