Merck
  • Determination of ofloxacin in tear by HPLC-ESI-MS/MS method: Comparison of ophthalmic drug release between a new mucoadhesive chitosan films and a conventional eye drop formulation in rabbit model
Determination of ofloxacin in tear by HPLC-ESI-MS/MS method: Comparison of ophthalmic drug release between a new mucoadhesive chitosan films and a conventional eye drop formulation in rabbit model
Journal of Pharmaceutical and Biomedical Analysis (2012)
Byrro RMD, et al.